Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7224UCPW
 View more versions of this product
Catalog No. 02-214-476
  • / Each
Description & Specifications
Specifications
Material Stainless Steel
Includes Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case
Classification UltraClass Platinum
Certifications/Compliance NVLAP, ISO9001
Quantity 9/Set
Weight (Metric) 1g to 100g
Product Type Medium metric analytical weight set
Mass 1 to 100g