Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7221UCPW
 View more versions of this product
Catalog No. 02-214-467
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Product Type Medium metric analytical weight set
Certifications/Compliance NVLAP, ISO9001
Includes Stainless Steel cylindrical weight: 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Classification UltraClass Platinum
Quantity 23/Set
Material Stainless Steel, Aluminum
Weight (Metric) 1mg to 200g
Mass 1mg to (2)200g