Troemner™ UltraClass Platinum Calibration Weight Sets with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
$5,707.74 - $9,887.97
Specifications
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Platinum
View More Specs
Products 25
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
02-214-455 View Documents Troemner™
7217UCPW
Stainless Steel, Aluminum 1mg to 1kg 1mg to 1kg 25/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-458 View Documents Troemner™
7218UCPW
Stainless Steel 1g to 500g 500g-1g 12/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-461 View Documents Troemner™
7219UCPW
Stainless Steel, Aluminum 1mg to 500g 1mg to 500g 24/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-464 View Documents Troemner™
7220UCPW
Stainless Steel 1g to 200g 2-200g-1g 11/Set Each for $9,551.89
02-214-467 View Documents Troemner™
7221UCPW
Stainless Steel, Aluminum 1mg to 200g 1mg to (2)200g 23/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-470 View Documents Troemner™
7222UCPW
Stainless Steel 1g to 200g 200g-1g 10/Set Each for $8,641.66
02-214-473 View Documents Troemner™
7223UCPW
Stainless Steel, Aluminum 1mg to 200g 1mg to 200g 22/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-476 View Documents Troemner™
7224UCPW
Stainless Steel 1g to 100g 1 to 100g 9/Set Each for $7,346.06
02-214-479 View Documents Troemner™
7225UCPW
Stainless Steel 100mg to 100g 100g-100mg 13/Set Each for $9,887.97
02-214-482 View Documents Troemner™
7226UCPW
Stainless Steel 10mg to 100g 10mg to 100g 17/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-485 View Documents Troemner™
7227UCPW
Stainless Steel 5mg to 100g 100g-5mg 18/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-488 View Documents Troemner™
7228UCPW
Stainless Steel, Aluminum 1mg to 100g 1mg to 100g 21/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-491 View Documents Troemner™
7229UCPW
Stainless Steel 1g to 50g 1 to 50g 8/Set Each for $6,742.14
02-214-494 View Documents Troemner™
7230UCPW
Stainless Steel 100mg to 50g 50g-100mg 12/Set Each for $9,317.40
02-214-497 View Documents Troemner™
7231UCPW
Stainless Steel 10mg to 50g 10mg to 50g 16/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-500 View Documents Troemner™
7232UCPW
Stainless Steel 5mg to 50g 50g-5mg 17/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-503 View Documents Troemner™
7233UCPW
Stainless Steel, Aluminum 1mg to 50g 50g-1mg 20/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-506 View Documents Troemner™
7234UCPW
Stainless Steel 1g to 20g 20g-1g 6/Set Each for $5,707.74
02-214-509 View Documents Troemner™
7235UCPW
Stainless Steel 100mg to 20g 20g-100mg 10/Set Each for $8,335.31
02-214-512 View Documents Troemner™
7236UCPW
Stainless Steel 10mg to 20g 20g-10mg 14/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-515 View Documents Troemner™
7237UCPW
Stainless Steel 5mg to 20g 20g-5mg 15/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-518 View Documents Troemner™
7238UCPW
Stainless Steel, Aluminum 1mg to 20g 20g-1mg 18/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-521 View Documents Troemner™
7239UCPW
Stainless Steel 10mg to 500mg 500mg-10mg 8/Set Each for $6,321.91
02-214-524 View Documents Troemner™
7240UCPW
Stainless Steel, Aluminum 1mg to 500g 1 to 500mg 12/Set Each for $7,341.46
02-100-155 View Documents Troemner™
7214UCPW
Stainless Steel 1g to 2kg 1g to 2kg 14/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Construction:
  • Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
  • Weights 3mg and below are made of aluminum
Note: Weight sets where the largest weight is 500g and below are supplied in a high-quality polycarbonate case. A wood case is provided for sets where the largest weight is 1kg and above.
Certifications
With NVLAP™ weight calibration certificate.
Specifications
Specifications
NVLAP, ISO9001
UltraClass Platinum