Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7240UCPW
 View more versions of this product
Catalog No. 02-214-524
  • / Each
Description & Specifications
Specifications
Product Type Small metric analytical weight set
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Platinum
Quantity 12/Set
Material Stainless Steel, Aluminum
Weight (Metric) 1mg to 500g
Mass 1 to 500mg
Includes Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case