Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7236UCPW
 View more versions of this product
Catalog No. 02-214-512
  • / Each
Description & Specifications
Specifications
Includes Cylindrical weight: 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case
Weight (Metric) 10mg to 20g
Product Type Medium metric analytical weight set
Classification UltraClass Platinum
Quantity 14/Set
Certifications/Compliance NVLAP, ISO9001
Mass 20g-10mg
Material Stainless Steel