Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7227UCPW
 View more versions of this product
Catalog No. 02-214-485
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Product Type Medium metric analytical weight set
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Platinum
Material Stainless Steel
Weight (Metric) 5mg to 100g
Mass 100g-5mg
Includes Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Polycarbonate case
Quantity 18/Set