Troemner™ UltraClass Platinum Calibration Weight Sets
For calibrating high-precision analytical balances with readability as low as 0.0001mg
$3,734.11 - $9,952.94
Specifications
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Platinum
View More Specs
Products 25
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
02-214-456 View Documents Troemner™
7218UCP
Stainless Steel 1g to 500g 500g-1g 12/Set Each for $9,952.94
02-214-462 View Documents Troemner™
7220UCP
Stainless Steel 1g to 200g 2-200g-1g 11/Set Each for $7,026.54
02-214-468 View Documents Troemner™
7222UCP
Stainless Steel 1g to 200g 200g-1g 10/Set Each for $6,033.71
02-214-477 View Documents Troemner™
7225UCP
Stainless Steel 100mg to 100g 100g-100mg 13/Set Each for $7,026.54
02-214-483 View Documents Troemner™
7227UCP
Stainless Steel 5mg to 100g 100g-5mg 18/Set Each for $8,780.71
02-214-492 View Documents Troemner™
7230UCP
Stainless Steel 100mg to 50g 50g-100mg 12/Set Each for $6,143.23
02-214-498 View Documents Troemner™
7232UCP
Stainless Steel 5mg to 50g 50g-5mg 17/Set Each for $8,194.60
02-214-501 View Documents Troemner™
7233UCP
Stainless Steel, Aluminum 1mg to 50g 50g-1mg 20/Set Each for $8,780.71
02-214-504 View Documents Troemner™
7234UCP
Stainless Steel 1g to 20g 20g-1g 6/Set Each for $4,333.26
02-214-507 View Documents Troemner™
7235UCP
Stainless Steel 100mg to 20g 20g-100mg 10/Set Each for $5,648.37
02-214-510 View Documents Troemner™
7236UCP
Stainless Steel 10mg to 20g 20g-10mg 14/Set Each for $7,201.23
02-214-513 View Documents Troemner™
7237UCP
Stainless Steel 5mg to 20g 20g-5mg 15/Set Each for $7,789.97
02-214-516 View Documents Troemner™
7238UCP
Stainless Steel, Aluminum 1mg to 20g 20g-1mg 18/Set Each for $7,962.57
02-214-519 View Documents Troemner™
7239UCP
Stainless Steel 10mg to 500mg 500mg-10mg 8/Set Each for $3,734.11
02-100-154 View Documents Troemner™
7214UCP
Stainless Steel 1g to 2kg 1g to 2kg 14/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-453 View Documents Troemner™
7217UCP
Stainless Steel, Aluminum 1mg to 1kg 1mg to 1kg 25/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-459 View Documents Troemner™
7219UCP
Stainless Steel, Aluminum 1mg to 500g 1mg to 500g 24/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-465 View Documents Troemner™
7221UCP
Stainless Steel, Aluminum 1mg to 200g 1mg to (2)200g 23/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-471 View Documents Troemner™
7223UCP
Stainless Steel, Aluminum 1mg to 200g 1mg to 200g 22/Set N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-474 View Documents Troemner™
7224UCP
Stainless Steel 1g to 100g 1 to 100g 9/Set Each for $4,424.00
02-214-480 View Documents Troemner™
7226UCP
Stainless Steel 10mg to 100g 10mg to 100g 17/Set Each for $7,091.74
02-214-486 View Documents Troemner™
7228UCP
Stainless Steel, Aluminum 1mg to 100g 1mg to 100g 21/Set Each for $9,279.74
02-214-489 View Documents Troemner™
7229UCP
Stainless Steel 1g to 50g 1 to 50g 8/Set Each for $4,780.66
02-214-495 View Documents Troemner™
7231UCP
Stainless Steel 10mg to 50g 10mg to 50g 16/Set Each for $7,502.63
02-214-522 View Documents Troemner™
7240UCP
Stainless Steel, Aluminum 1mg to 500g 1 to 500mg 12/Set Each for $5,270.83
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Construction:
  • Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
  • Weights 3mg and below are made of aluminum
Note: Weight sets where the largest weight is 500g and below are supplied in a high-quality polycarbonate case. A wood case is provided for sets where the largest weight is 1kg and above.
Certifications
Includes Statement of Accuracy.
Specifications
Specifications
Statement of accuracy, ISO9001
UltraClass Platinum