Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7239UCP
 View more versions of this product
Catalog No. 02-214-519
  • / Each
Description & Specifications
Specifications
Product Type Small metric analytical weight set
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Platinum
Material Stainless Steel
Weight (Metric) 10mg to 500mg
Mass 500mg-10mg
Includes Sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case
Quantity 8/Set