Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7220UCP
 View more versions of this product
Catalog No. 02-214-462
  • / Each
Description & Specifications
Specifications
Product Type Medium metric analytical weight set
Certifications/Compliance Statement of accuracy, ISO9001
Weight (Metric) 1g to 200g
Material Stainless Steel
Quantity 11/Set
Classification UltraClass Platinum
Includes Cylindrical weight: 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case
Mass 2-200g-1g