Please login to your online account to display your discounted pricing
Troemner™ UltraClass Platinum Calibration Weight Sets
For calibrating high-precision analytical balances with readability as low as 0.0001mg
Manufacturer: troemner llc  7225UCP
 View more versions of this product
Catalog No. 02-214-477
  • / Each
Description & Specifications
Specifications
Classification UltraClass Platinum
Quantity 13/Set
Includes Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, Polycarbonate case
Weight (Metric) 100mg to 100g
Mass 100g-100mg
Product Type Medium metric analytical weight set
Certifications/Compliance Statement of accuracy, ISO9001
Material Stainless Steel