Learn More
Troemner™ UltraClass Gold Individual Analytical Precision Weights with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as .001mg
$662.26 - $6,187.06
Specifications
Certifications/Compliance | Traceable certificate, ISO9001 |
---|---|
Classification | UltraClass Gold |
Includes | Polycarbonate case |
Quantity | Each |
Catalog Number | Mfr. No. | Material | Mass | Shape | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Mass | Shape | Price | Quantity | |||
02-214-215
|
Troemner™
7026UCGT |
Stainless Steel | 500 mg | Flat with one end turned up for easy handling |
Each for $696.03
|
|
02-214-218
|
Troemner™
7014UCGT |
Stainless Steel | 500 g | Cylindrical with Handling Knob |
Each for $1,663.80
|
|
02-214-221
|
Troemner™
7030UCGT |
Stainless Steel | 50 mg | Flat with one end turned up for easy handling |
Each for $747.40
|
|
02-214-227
|
Troemner™
7034UCGT |
Stainless Steel | 5 mg | Flat with one end turned up for easy handling |
Each for $696.03
|
|
02-214-230
|
Troemner™
7010UCGT |
Stainless Steel | 5 kg | Cylindrical with Handling Knob |
Each for $6,187.06
|
|
02-214-233
|
Troemner™
7022UCGT |
Stainless Steel | 5 g | Cylindrical with Handling Knob |
Each for $813.46
|
|
02-214-236
|
Troemner™
7027UCGT |
Stainless Steel | 300 mg | Flat with one end turned up for easy handling |
Each for $711.14
|
|
02-214-239
|
Troemner™
7015UCGT |
Stainless Steel | 300 g | Cylindrical with Handling Knob |
Each for $1,362.43
|
|
02-214-242
|
Troemner™
7031UCGT |
Aluminum | 30 mg | Flat with one end turned up for easy handling |
Each for $711.14
|
|
02-214-245
|
Troemner™
7019UCGT |
Stainless Steel | 30 g | Cylindrical with Handling Knob |
Each for $830.83
|
|
02-214-248
|
Troemner™
7035UCGT |
Aluminum | 3 mg | Flat with one end turned up for easy handling |
Each for $711.14
|
|
02-214-251
|
Troemner™
7011UCGT |
Stainless Steel | 3 kg | Cylindrical with Handling Knob |
Each for $5,178.43
|
|
02-214-254
|
Troemner™
7023UCGT |
Stainless Steel | 3 g | Cylindrical with Handling Knob |
Each for $830.83
|
|
02-214-257
|
Troemner™
7028UCGT |
Stainless Steel | 200 mg | Flat with one end turned up for easy handling |
Each for $662.26
|
|
02-214-260
|
Troemner™
7016UCGT |
Stainless Steel | 200 g | Cylindrical with Handling Knob |
Each for $1,233.77
|
|
02-214-263
|
Troemner™
7032UCGT |
Stainless Steel | 20 mg | Flat with one end turned up for easy handling |
Each for $662.26
|
|
02-214-269
|
Troemner™
7036UCGT |
Aluminum | 2 mg | Flat with one end turned up for easy handling |
Each for $662.26
|
|
02-214-272
|
Troemner™
7012UCGT |
Stainless Steel | 2 kg | Cylindrical with Handling Knob |
Each for $3,968.60
|
|
02-214-275
|
Troemner™
7024UCGT |
Stainless Steel | 2 g | Cylindrical with Handling Knob |
Each for $773.97
|
|
02-214-278
|
Troemner™
7029UCGT |
Stainless Steel | 100 mg | Flat with one end turned up for easy handling |
Each for $711.14
|
|
02-214-290
|
Troemner™
7037UCGT |
Aluminum | 1 mg | Flat with one end turned up for easy handling |
Each for $711.14
|
|
02-214-293
|
Troemner™
7013UCGT |
Stainless Steel | 1 kg | Cylindrical with Handling Knob |
Each for $2,780.20
|
|
02-214-296
|
Troemner™
7025UCGT |
Stainless Steel | 1 g | Cylindrical with Handling Knob |
Each for $830.83
|
|
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
- Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
- This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
- Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
- Weights come with a Positive Calibration Tolerance Guarantee
Certifications
With Traceable certificate.
Specifications
Traceable certificate, ISO9001 | |
UltraClass Gold | |
Polycarbonate case | |
Each |