Troemner™ UltraClass Gold Individual Analytical Precision Weights with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as .001mg
Manufacturer:  Troemner™ 7017UCGT
 View more versions of this product
Catalog No. 02-214-281
Description
Description
Certifications
With Traceable certificate.
Specifications
Specifications
Metric analytical weight with handling knob
UltraClass Gold
100g
100g
Polycarbonate case
Traceable certificate, ISO9001
Stainless Steel
8.03g/cm3 at 20°C
Cylindrical with handling knob
Each
Documents