Please login to your online account to display your discounted pricing

Troemner™ UltraClass Gold Individual Analytical Precision Weights with NVLAP Certificate


For calibrating high-precision analytical balances with readability as low as .001mg

Manufacturer: troemner llc  7037UCGW

 View more versions of this product

Catalog No. 02-214-288

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance NVLAP, ISO9001
Includes Polycarbonate case
Density 2.7g/cm3 at 20°C
Weight (Metric) 1mg
Quantity Each
Classification UltraClass Gold
Shape Flat with one end turned up for easy handling
Mass 1mg
Product Type Metric analytical sheet weight with one side turned up
Material 3003-H14 Aluminum