Please login to your online account to display your discounted pricing

Troemner™ UltraClass Gold Individual Analytical Precision Weights with NVLAP Certificate


For calibrating high-precision analytical balances with readability as low as .001mg

Manufacturer: troemner llc  7036UCGW

 View more versions of this product

Catalog No. 02-214-267

  • / Each
 

Description & Specifications

Specifications

Product Type Metric analytical sheet weight with one side turned up
Certifications/Compliance NVLAP, ISO9001
Includes Polycarbonate case
Classification UltraClass Gold
Quantity Each
Material 3003-H14 Aluminum
Weight (Metric) 2mg
Density 2.7g/cm3 at 20°C
Mass 2mg
Shape Flat with one end turned up for easy handling