Troemner™ UltraClass Gold Individual Analytical Precision Weights
For calibrating high-precision analytical balances with readability as low as .001mg
$562.80 - $5,842.57
Specifications
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Gold
Includes Polycarbonate case
Quantity Each
View More Specs
Products 18
Catalog Number Mfr. No. Material Mass Shape Price Quantity  
Catalog Number Mfr. No. Material Mass Shape Price Quantity  
02-214-217
View Documents
Troemner™
7014UCG
Stainless Steel 500 g Cylindrical with Handling Knob
Each for $1,432.34
02-214-220
View Documents
Troemner™
7030UCG
Stainless Steel 50 mg Flat with one end turned up for easy handling
Each for $591.51
02-214-229
View Documents
Troemner™
7010UCG
Stainless Steel 5 kg Cylindrical with Handling Knob
Each for $5,842.57
02-214-235
View Documents
Troemner™
7027UCG
Stainless Steel 300 mg Flat with one end turned up for easy handling
Each for $562.80
02-214-238
View Documents
Troemner™
7015UCG
Stainless Steel 300 g Cylindrical with Handling Knob
Each for $1,189.03
02-214-244
View Documents
Troemner™
7019UCG
Stainless Steel 30 g Cylindrical with Handling Knob
Each for $616.83
02-214-241
View Documents
Troemner™
7031UCG
Aluminum 30 mg Flat with one end turned up for easy handling
Each for $562.80
02-214-247
View Documents
Troemner™
7035UCG
Aluminum 3 mg Flat with one end turned up for easy handling
Each for $562.80
02-214-253
View Documents
Troemner™
7023UCG
Stainless Steel 3 g Cylindrical with Handling Knob
Each for $616.83
02-214-265
View Documents
Troemner™
7020UCG
Stainless Steel 20 g Cylindrical with Handling Knob
Each for $616.83
02-214-268
View Documents
Troemner™
7036UCG
Aluminum 2 mg Flat with one end turned up for easy handling
Each for $562.80
02-214-271
View Documents
Troemner™
7012UCG
Stainless Steel 2 kg Cylindrical with Handling Knob
Each for $3,615.91
02-214-277
View Documents
Troemner™
7029UCG
Stainless Steel 100 mg Flat with one end turned up for easy handling
Each for $562.80
02-214-274
View Documents
Troemner™
7024UCG
Stainless Steel 2 g Cylindrical with Handling Knob
Each for $616.83
02-214-280
View Documents
Troemner™
7017UCG
Stainless Steel 100 g Cylindrical with Handling Knob
Each for $760.11
02-214-283
View Documents
Troemner™
7033UCG
Stainless Steel 10 mg Flat with one end turned up for easy handling
Each for $562.80
02-214-286
View Documents
Troemner™
7021UCG
Stainless Steel 10 g Cylindrical with Handling Knob
Each for $616.83
02-214-295
View Documents
Troemner™
7025UCG
Stainless Steel 1 g Cylindrical with Handling Knob
Each for $616.83
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Note: Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum). Weights 3mg and below are made of aluminum.
Certifications
Includes Statement of Accuracy.
Specifications
Specifications
Statement of accuracy, ISO9001
UltraClass Gold
Polycarbonate case
Each