missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Troemner™ UltraClass Gold Individual Analytical Precision Weights
For calibrating high-precision analytical balances with readability as low as .001mg
Manufacturer: Troemner™ 7036UCG
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
- Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
- This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
- Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
- Weights come with a Positive Calibration Tolerance Guarantee
Certifications
Includes Statement of Accuracy.
Specifications
Metric Analytical Sheet Weight with One Side Turned Up | |
UltraClass Gold | |
2 mg | |
2 mg | |
Polycarbonate case |
Statement of accuracy, ISO9001 | |
Aluminum | |
2.7g/cm3 at 20°C | |
Flat with one end turned up for easy handling | |
Each |