Troemner™ UltraClass Gold Electronic Balance Calibration Weights with Traceable Certificate
Constructed of special Troemner Alloy 8 stainless steel
Manufacturer:  Troemner Llc 8024UCGT
 View more versions of this product
Catalog No. 02-218-170
Specifications Description & Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
UltraClass Gold
2kg
2kg
Polypropylene case
Traceable certificate, ISO9001
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with groove handle
Each