Troemner™ UltraClass Gold Electronic Balance Calibration Weights with Traceable Certificate
Constructed of special Troemner Alloy 8 stainless steel
Manufacturer:  Henry Troemner Inc 8032UCGT
 View more versions of this product
Catalog No. 02-218-167
Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
UltraClass Gold
500g
500g
Polypropylene case
Traceable certificate, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Straight cylinder
Each
Documents