Troemner™ UltraClass Gold Electronic Balance Calibration Weights with Traceable Certificate
Constructed of special Troemner Alloy 8 stainless steel
$345.09 - $7,929.71
Specifications
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass Gold
Product Type Two-Piece Electronic Balance Calibration Weight
Quantity Each
View More Specs
Products 16
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
02-218-167 View Documents Henry Troemner Inc
8032UCGT
Polypropylene case 500g Straight cylinder Each for $657.46
02-218-172 View Documents Henry Troemner Inc
8016UCGT
Polypropylene case 5kg Cylindrical with groove handle Each for $1,746.97
02-218-176 View Documents Henry Troemner Inc
8010UCGT
Without case 10kg Cylindrical with recessed grip handle Each for $3,608.80
02-218-145 View Documents Henry Troemner Inc
8054UCGT
Polypropylene case 30g Straight cylinder Each for $345.09
02-218-148 View Documents Henry Troemner Inc
8053UCGT
Polypropylene case 40g Straight cylinder Each for $356.00
02-218-151 View Documents Henry Troemner Inc
8052UCGT
Polypropylene case 50g Straight cylinder Each for $345.09
02-218-153 View Documents Henry Troemner Inc
8044UCGT
Polypropylene case 100g Straight cylinder Each for $388.69
02-218-156 View Documents Henry Troemner Inc
8042UCGT
Polypropylene case 150g Straight cylinder Each for $504.91
02-218-159 View Documents Henry Troemner Inc
8040UCGT
Polypropylene case 160g Straight cylinder Each for $504.91
02-218-162 View Documents Henry Troemner Inc
8036UCGT
Polypropylene case 300g Straight cylinder Each for $566.66
02-218-165 View Documents Henry Troemner Inc
8034UCGT
Polypropylene case 400g Straight cylinder Each for $632.03
02-218-170 View Documents Henry Troemner Inc
8024UCGT
Polypropylene case 2kg Cylindrical with groove handle Each for $929.91
02-218-174 View Documents Henry Troemner Inc
8014UCGT
Polypropylene case 5kg Cylindrical with recessed grip handle Each for $2,709.86
02-218-179 View Documents Henry Troemner Inc
8006UCGT
Without case 20kg Cylindrical with recessed grip handle Each for $5,459.60
02-218-182 View Documents Henry Troemner Inc
8002UCGT
Without case 25kg Cylindrical with recessed grip handle Each for $6,658.34
02-218-185 View Documents Henry Troemner Inc
8000UCGT
Without case 30kg Cylindrical with recessed grip handle Each for $7,929.71
Description
Description
UltraClass Gold Series Weights combine high precision with the advantage of two-piece construction, avoiding costly replacement issues associated with one-piece weights. Consistent, automated robotic measurement means that Troemner UltraClass Series weights are the most precise two-piece weights with tolerances that are equal to ANSI/ASTM E617 Class 000 (UltraClass Platinum), Class 00 (UltraClass Gold) and Class 0 (UltraClass).
Two-piece weights with adjusting cavities
  • Made from stainless steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properties
  • UltraClass™ weights are highly polished to a mirror-like finish
Precision-Machined Stainless-Steel in Three Styles
  • Masses from 30 to 500g: straight cylinders
  • 1 to 5kg: cylindrical with indentation near top for easy gripping
  • 5 to 30kg: sleek cylinders with recessed grip handles, flat on top, easily stacked on center
Protective Cases
  • Individual weights 10g through 5kg supplied in high-quality polycarbonate case
  • Soft absorbent cotton glove is included to protect weights during use
  • Optional cases available for grip-handle weights
Certifications
With Traceable Certificate.
Specifications
Specifications
Traceable certificate, ISO9001
UltraClass Gold
Two-Piece Electronic Balance Calibration Weight
Each