Troemner™ UltraClass Gold Electronic Balance Calibration Weights with NVLAP Certificate
Constructed of special Troemner Alloy 8 stainless steel
Manufacturer:  Henry Troemner Inc 8054UCGW
 View more versions of this product
Catalog No. 02-218-144
Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
UltraClass Gold
30g
30g
Polypropylene case
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Straight cylinder
Each
Documents