Troemner™ UltraClass Gold Electronic Balance Calibration Weights with NVLAP Certificate
Constructed of special Troemner Alloy 8 stainless steel
Manufacturer:  Henry Troemner Inc 8002UCGW
 View more versions of this product
Catalog No. 02-218-181
Specifications
Specifications
Electronic balance calibration weight with grip handle
UltraClass Gold
25kg
25kg
Without case
NVLAP, ISO9001
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with recessed grip handle
Each
Documents