Please login to your online account to display your discounted pricing
Troemner™ UltraClass Gold Electronic Balance Calibration Weights with NVLAP Certificate
Constructed of special Troemner Alloy 8 stainless steel
$518.86 - $8,706.89
Specifications
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Gold
Product Type Two piece electronic balance calibration weight
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Includes Mass Shape Price Quantity    
02-218-144
troemner llc
8054UCGW
Polypropylene case 30g Straight cylinder Each for $518.86
02-218-147
troemner llc
8053UCGW
Polypropylene case 40g Straight cylinder Each for $518.86
02-218-150
troemner llc
8052UCGW
Polypropylene case 50g Straight cylinder Each for $518.86
02-218-155
troemner llc
8042UCGW
Polypropylene case 150g Straight cylinder Each for $806.71
02-218-158
troemner llc
8040UCGW
Polypropylene case 160g Straight cylinder Each for $806.71
02-218-164
troemner llc
8034UCGW
Polypropylene case 400g Straight cylinder Each for $806.71
02-218-173
troemner llc
8014UCGW
Polypropylene case 5kg Cylindrical with recessed grip handle Each for $2,860.83
02-218-178
troemner llc
8006UCGW
Without case 20kg Cylindrical with recessed grip handle Each for $5,760.77
02-218-181
troemner llc
8002UCGW
Without case 25kg Cylindrical with recessed grip handle Each for $6,929.97
02-218-184
troemner llc
8000UCGW
Without case 30kg Cylindrical with recessed grip handle Each for $8,706.89
Description & Specifications
Specifications
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Gold
Product Type Two piece electronic balance calibration weight
Quantity Each
UltraClass Gold Series Weights combine high precision with the advantage of two-piece construction, avoiding costly replacement issues associated with one-piece weights. Consistent, automated robotic measurement means that Troemner UltraClass Series weights are the most precise two-piece weights with tolerances that are equal to ANSI/ASTM E617 Class 000 (UltraClass Platinum), Class 00 (UltraClass Gold) and Class 0 (UltraClass).
Two-piece weights with adjusting cavities
  • Made from stainless steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properties
  • UltraClass™ weights are highly polished to a mirror-like finish
Precision-Machined Stainless-Steel in Three Styles
  • Masses from 30 to 500g: straight cylinders
  • 1 to 5kg: cylindrical with indentation near top for easy gripping
  • 5 to 30kg: sleek cylinders with recessed grip handles, flat on top, easily stacked on center
Protective Cases
  • Individual weights 10g through 5kg supplied in high-quality polycarbonate case
  • Soft absorbent cotton glove is included to protect weights during use
  • Optional cases available for grip-handle weights
With NIST™/NVLAP™ Weight Calibration Certificate