Troemner™ UltraClass Gold Electronic Balance Calibration Weights
Constructed of special Troemner Alloy 8 stainless steel
$232.31 - $7,355.71
Specifications
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Gold
Product Type Two-Piece Electronic Balance Calibration Weight
Quantity Each
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
Catalog Number Mfr. No. Includes Mass Shape Price Quantity  
02-218-161 Troemner Llc
8036UCG
Polypropylene case 300g Straight cylinder Each for $411.09
02-218-166 Troemner Llc
8032UCG
Polypropylene case 500g Straight cylinder Each for $510.23
02-218-168 Troemner Llc
8028UCG
Polypropylene case 1kg Cylindrical with groove handle Each for $613.14
02-218-143 Troemner Llc
8054UCG
Polypropylene case 30g Straight cylinder Each for $232.31
02-218-149 Troemner Llc
8052UCG
Polypropylene case 50g Straight cylinder Each for $246.63
02-218-152 Troemner Llc
8044UCG
Polypropylene case 100g Straight cylinder Each for $300.23
02-218-154 Troemner Llc
8042UCG
Polypropylene case 150g Straight cylinder Each for $375.31
02-218-157 Troemner Llc
8040UCG
Polypropylene case 160g Straight cylinder Each for $375.31
02-218-160 Troemner Llc
8038UCG
Polypropylene case 200g Straight cylinder Each for $361.00
02-218-163 Troemner Llc
8034UCG
Polypropylene case 400g Straight cylinder Each for $486.06
02-218-169 Troemner Llc
8024UCG
Polypropylene case 2kg Cylindrical with groove handle Each for $718.40
02-218-171 Troemner Llc
8016UCG
Polypropylene case 5kg Cylindrical with groove handle Each for $1,622.69
02-218-175 Troemner Llc
8010UCG
Without case 10kg Cylindrical with recessed grip handle Each for $3,491.97
02-218-177 Troemner Llc
8006UCG
Without case 20kg Cylindrical with recessed grip handle Each for $5,186.17
02-218-180 Troemner Llc
8002UCG
Without case 25kg Cylindrical with recessed grip handle Each for $6,229.83
02-218-183 Troemner Llc
8000UCG
Without case 30kg Cylindrical with recessed grip handle Each for $7,355.71
Specifications
Description
UltraClass Gold Series Weights combine high precision with the advantage of two-piece construction, avoiding costly replacement issues associated with one-piece weights. Consistent, automated robotic measurement means that Troemner UltraClass Series weights are the most precise two-piece weights with tolerances that are equal to ANSI/ASTM E617 Class 000 (UltraClass Platinum), Class 00 (UltraClass Gold) and Class 0 (UltraClass).
Two-piece weights with adjusting cavities
  • Made from stainless steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properties
  • UltraClass™ weights are highly polished to a mirror-like finish
Precision-Machined Stainless-Steel in Three Styles
  • Masses from 30 to 500g: straight cylinders
  • 1 to 5kg: cylindrical with indentation near top for easy gripping
  • 5 to 30kg: sleek cylinders with recessed grip handles, flat on top, easily stacked on center
Protective Cases
  • Individual weights 10g through 5kg supplied in high-quality polycarbonate case
  • Soft absorbent cotton glove is included to protect weights during use
  • Optional cases available for grip-handle weights
Description & Specifications
Specifications
Statement of accuracy, ISO9001
UltraClass Gold
Two-Piece Electronic Balance Calibration Weight
Each