Learn More
Troemner™ UltraClass Gold Calibration Weight Sets with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as .001mg
$3,772.11 - $9,680.63
Specifications
Certifications/Compliance | Traceable certificate, ISO9001 |
---|---|
Classification | UltraClass Gold |
Catalog Number | Mfr. No. | Material | Weight (Metric) | Mass | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Weight (Metric) | Mass | Quantity | Price | Quantity | ||
02-214-298
|
Troemner™
7217UCGT |
Stainless Steel, Aluminum | 1mg to 1kg | 1 kg to 1 mg | 25/Set |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
02-214-304
|
Troemner™
7219UCGT |
Stainless Steel, Aluminum | 1 mg to 500 g | 500 g to 1 mg | 24/Set |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
02-214-301
|
Troemner™
7218UCGT |
Stainless Steel | 1 g to 500 g | 500 g to 1 g | 12/Set |
Each for $9,622.77
|
|
02-214-307
|
Troemner™
7220UCGT |
Stainless Steel | 1 g to 200 g | 2 to 200 g to 1 g | 11/Set |
Each for $6,356.34
|
|
02-214-310
|
Troemner™
7221UCGT |
Stainless Steel, Aluminum | 1 mg to 200 g | 2 to 200 g to 1 mg | 23/Set |
Each for $9,680.63
|
|
02-214-322
|
Troemner™
7225UCGT |
Stainless Steel | 100 mg to 100 g | 100 g to 100 mg | 13/Set |
Each for $6,245.11
|
|
02-214-331
|
Troemner™
7228UCGT |
Stainless Steel, Aluminum | 1 mg to 100 g | 100 g to 1 mg | 21/Set |
Each for $8,213.46
|
|
02-214-328
|
Troemner™
7227UCGT |
Stainless Steel | 5 mg to 100 g | 100 g to 5 mg | 18/Set |
Each for $7,307.34
|
|
02-214-334
|
Troemner™
7229UCGT |
Stainless Steel | 1 g to 50 g | 50 g to 1 g | 8/Set |
Each for $4,077.80
|
|
02-214-337
|
Troemner™
7230UCGT |
Stainless Steel | 100 mg to 50 g | 50 g to 100 mg | 12/Set |
Each for $5,382.40
|
|
02-214-340
|
Troemner™
7231UCGT |
Stainless Steel | 10 mg to 50 g | 50 g to 10 mg | 16/Set |
Each for $7,654.43
|
|
02-214-346
|
Troemner™
7233UCGT |
Stainless Steel, Aluminum | 1 mg to 50 g | 50 g to 1 mg | 20/Set |
Each for $7,481.40
|
|
02-214-343
|
Troemner™
7232UCGT |
Stainless Steel | 5 mg to 50 g | 50 g to 5 mg | 17/Set |
Each for $6,577.26
|
|
02-214-349
|
Troemner™
7234UCGT |
Stainless Steel | 1 g to 20 g | 20 g to 1 g | 6/Set |
Each for $3,795.06
|
|
02-214-352
|
Troemner™
7235UCGT |
Stainless Steel | 100 mg to 20 g | 20 g to 100 mg | 10/Set |
Each for $4,765.51
|
|
02-214-355
|
Troemner™
7236UCGT |
Stainless Steel | 10 mg to 20 g | 20 g to 10 mg | 14/Set |
Each for $5,844.69
|
|
02-214-358
|
Troemner™
7237UCGT |
Stainless Steel | 5 mg to 20 g | 20 g to 5 mg | 15/Set |
Each for $5,362.43
|
|
02-214-364
|
Troemner™
7239UCGT |
Stainless Steel | 10mg to 500mg | 500 mg to 10 mg | 8/Set |
Each for $3,772.11
|
|
02-214-367
|
Troemner™
7240UCGT |
Stainless Steel, Aluminum | 1 mg to 500 g | 500 mg to 1 mg | 12/Set |
Each for $4,143.14
|
|
02-100-160
|
Troemner™
7214UCGT |
Stainless Steel | 1 g to 2 kg | 1 g to 2 kg | 14/Set |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
- Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
- This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
- Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
- Weights come with a Positive Calibration Tolerance Guarantee
- Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
- Weights 3mg and below are made of aluminum
Certifications
With Traceable certificate.
Specifications
Traceable certificate, ISO9001 | |
UltraClass Gold |