Please login to your online account to display your discounted pricing
Troemner™ UltraClass Gold Calibration Weight Sets with Traceable Certificate
For calibrating high-precision analytical balances with readability as low as .001mg
Manufacturer: troemner llc  7238UCGT
 View more versions of this product
Catalog No. 02-214-361
  • / Each
Description & Specifications
Specifications
Quantity 18/Set
Weight (Metric) 1mg to 20g
Product Type Medium metric analytical weight set
Classification UltraClass Gold
Includes Stainless Steel cylindrical weight: 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Certifications/Compliance Traceable certificate, ISO9001
Material Stainless Steel, Aluminum
Mass 20g-1mg