Troemner™ UltraClass Gold Calibration Weight Sets with NVLAP Certificate
For calibrating high-precision analytical balances with readability as low as .001mg
$5,665.26 - $5,665.26
Specifications
Certifications/Compliance NVLAP, ISO9001
Classification UltraClass Gold
View More Specs
Products 5
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
02-214-299
View Documents
Troemner™
7217UCGW
Stainless Steel, Aluminum 1mg to 1kg 1 kg to 1 mg 25/Set N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-305
View Documents
Troemner™
7219UCGW
Stainless Steel, Aluminum 1 mg to 500 g 500 g to 1 mg 24/Set N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-311
View Documents
Troemner™
7221UCGW
Stainless Steel, Aluminum 1 mg to 200 g 2 to 200 g to 1 mg 23/Set N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-368
View Documents
Troemner™
7240UCGW
Stainless Steel, Aluminum 1 mg to 500 g 500 mg to 1 mg 12/Set
Each for $5,665.26
02-100-159
View Documents
Troemner™
7214UCGW
Stainless Steel 1 g to 2 kg 1 g to 2 kg 14/Set N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Construction:
  • Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
  • Weights 3mg and below are made of aluminum
Note: Weight sets where the largest weight is 500g and below are supplied in a high-quality polycarbonate case. A wood case is provided for sets where the largest weight is 1kg and above.
Certifications
With NVLAP™ weight calibration certificate.
Specifications
Specifications
NVLAP, ISO9001
UltraClass Gold