Troemner™ UltraClass Gold Calibration Weight Sets
For calibrating high-precision analytical balances with readability as low as .001mg
$2,835.11 - $8,701.37
Specifications
Certifications/Compliance Statement of accuracy, ISO9001
Classification UltraClass Gold
View More Specs
Products 23
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
Catalog Number Mfr. No. Material Weight (Metric) Mass Quantity Price Quantity  
02-214-297
View Documents
Troemner™
7217UCG
Stainless Steel, Aluminum 1mg to 1kg 1 kg to 1 mg 25/Set N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-300
View Documents
Troemner™
7218UCG
Stainless Steel 1 g to 500 g 500 g to 1 g 12/Set
Each for $7,556.34
02-214-303
View Documents
Troemner™
7219UCG
Stainless Steel, Aluminum 1 mg to 500 g 500 g to 1 mg 24/Set N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02-214-306
View Documents
Troemner™
7220UCG
Stainless Steel 1 g to 200 g 2 to 200 g to 1 g 11/Set
Each for $5,337.54
02-214-309
View Documents
Troemner™
7221UCG
Stainless Steel, Aluminum 1 mg to 200 g 2 to 200 g to 1 mg 23/Set
Each for $8,701.37
02-214-312
View Documents
Troemner™
7222UCG
Stainless Steel 1 g to 200 g 200 g to 1 g 10/Set
Each for $4,583.11
02-214-315
View Documents
Troemner™
7223UCG
Stainless Steel, Aluminum 1 mg to 200 g 200 g to 1 mg 22/Set
Each for $7,770.20
02-214-318
View Documents
Troemner™
7224UCG
Stainless Steel 1 g to 100 g 100 g to 1 g 9/Set
Each for $3,360.34
02-214-321
View Documents
Troemner™
7225UCG
Stainless Steel 100 mg to 100 g 100 g to 100 mg 13/Set
Each for $5,337.54
02-214-324
View Documents
Troemner™
7226UCG
Stainless Steel 10 mg to 100 g 100 g to 10 mg 17/Set
Each for $5,384.09
02-214-330
View Documents
Troemner™
7228UCG
Stainless Steel, Aluminum 1 mg to 100 g 100 g to 1 mg 21/Set
Each for $7,556.34
02-214-333
View Documents
Troemner™
7229UCG
Stainless Steel 1 g to 50 g 50 g to 1 g 8/Set
Each for $3,629.66
02-214-336
View Documents
Troemner™
7230UCG
Stainless Steel 100 mg to 50 g 50 g to 100 mg 12/Set
Each for $4,668.26
02-214-339
View Documents
Troemner™
7231UCG
Stainless Steel 10 mg to 50 g 50 g to 10 mg 16/Set
Each for $5,696.94
02-214-342
View Documents
Troemner™
7232UCG
Stainless Steel 5 mg to 50 g 50 g to 5 mg 17/Set
Each for $6,223.17
02-214-345
View Documents
Troemner™
7233UCG
Stainless Steel, Aluminum 1 mg to 50 g 50 g to 1 mg 20/Set
Each for $6,668.71
02-214-348
View Documents
Troemner™
7234UCG
Stainless Steel 1 g to 20 g 20 g to 1 g 6/Set
Each for $3,290.06
02-214-351
View Documents
Troemner™
7235UCG
Stainless Steel 100 mg to 20 g 20 g to 100 mg 10/Set
Each for $4,290.03
02-214-354
View Documents
Troemner™
7236UCG
Stainless Steel 10 mg to 20 g 20 g to 10 mg 14/Set
Each for $5,468.74
02-214-357
View Documents
Troemner™
7237UCG
Stainless Steel 5 mg to 20 g 20 g to 5 mg 15/Set
Each for $5,916.74
02-214-363
View Documents
Troemner™
7239UCG
Stainless Steel 10mg to 500mg 500 mg to 10 mg 8/Set
Each for $2,835.11
02-214-366
View Documents
Troemner™
7240UCG
Stainless Steel, Aluminum 1 mg to 500 g 500 mg to 1 mg 12/Set
Each for $4,001.91
02-100-158
View Documents
Troemner™
7214UCG
Stainless Steel 1 g to 2 kg 1 g to 2 kg 14/Set N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
  • Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
  • This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
  • Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
  • Weights come with a Positive Calibration Tolerance Guarantee
Construction:
  • Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
  • Weights 3mg and below are made of aluminum
Note: Weight sets where the largest weight is 500g and below are supplied in a high-quality polycarbonate case. A wood case is provided for sets where the largest weight is 1kg and above.
Certifications
Includes Statement of Accuracy.
Specifications
Specifications
Statement of accuracy, ISO9001
UltraClass Gold