missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Troemner™ UltraClass Gold Calibration Weight Sets
For calibrating high-precision analytical balances with readability as low as .001mg
Manufacturer: Troemner™ 7220UCG
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage. Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F).
- Weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1
- This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured
- Appropriate for calibrating high-precision analytical balances with a readability as low as .001mg
- Weights come with a Positive Calibration Tolerance Guarantee
- Fractional weights 5 to 500mg are made of stainless steel (except 30mg, which are aluminum)
- Weights 3mg and below are made of aluminum
Certifications
Includes Statement of Accuracy.
Specifications
Medium Metric Analytical Weight Set | |
UltraClass Gold | |
1 g to 200 g | |
Cylindrical weight: 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case |
Statement of accuracy, ISO9001 | |
Stainless Steel | |
2 to 200 g to 1 g | |
11/Set |