Troemner™ Talboys™ Tube Hosecocks
Flow control devices deliver accurate regulation or interruption of fluid flow.
Manufacturer:  Troemner Llc 916184
 View more versions of this product
Catalog No. 02-300-236
Specifications
Description
Hosecock, Tube; Troemner Talboys; Heavy duty; Nicke plated; One-hand operation; Convex bearing surface; Rounded edges; Adjustment screw; Built-in side lugs for foot mounting; For tubing 0-29mm; 57W x 106mm H
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
106mm
Nickel-plated zinc
1.14 in.
Adjustment screw with oversized head
Accurate regulation
Heavy Duty Hosecock
29mm