missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Troemner™ Talboys™ Support Plates
Holds hotplates, stirrers, beakers, mantles and other apparatus on lab frames and ring stands
Manufacturer: Troemner™ 916206
Description
- Constructed of durable aluminum
- Available in square or round shape
- Square plates include a non-skid rubber mat with built in holder that grips rods up to 19mm (0.75 in.)
Specifications
Support Plate | |
Round | |
15.2cm | |
6.6cm | |
6 in. (152mm) |
Aluminum, Rubber | |
6 in. | |
2.6 in. | |
Each |