Please login to your online account to display your discounted pricing
Troemner™ Talboys™ Labjaws™ Bench Clamps
Durable, cast-aluminum clamps secure gas cylinders to benches, tables, or other flat surfaces
Manufacturer: troemner llc  972036
 View more versions of this product
Catalog No. 02-216-363
  • / Each
Description & Specifications
Specifications
Product Type Bench Clamp w/Strap and Chain
Includes 54 L x 1 in. W Nylon strap with a Nickel plated, non-slip spring catch and buckle for simple adjustment around cylinders, strap and chain
Length (English) 3.25 in.
Length (Metric) 8.25cm
Material Cast Aluminum