Troemner™ Stainless Steel Electronic Balance Calibration Weights, UltraClass with Statement of Accuracy
Precision-crafted of stainless steel, with a highly polished finish
Manufacturer:  Troemner™ 8016*
 View more versions of this product
Catalog No. 02-214-6F
Description
Description
Order Info
  • Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove
  • Standard weights are supplied with a Statement of Accuracy
Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
UltraClass
5kg
5kg
Polypropylene case
1
Statement of accuracy, ISO9001
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with groove
Each
Documents
Accessories