Troemner™ Stainless Steel Electronic Balance Calibration Weights, UltraClass with Statement of Accuracy
Precision-crafted of stainless steel, with a highly polished finish
Manufacturer:  Troemner Llc 8024
 View more versions of this product
Catalog No. 02-214-6G
Specifications Description & Specifications
Specifications
Two piece electronic balance calibration weight
UltraClass
2kg
2kg
Polypropylene case
1
Statement of accuracy, ISO9001
303 Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with groove
Each
Accessories