Please login to your online account to display your discounted pricing

Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 4 with Statement of Accuracy


Precision-crafted of stainless steel, with a highly polished finish

Manufacturer: troemner llc  8452F

 View more versions of this product

Catalog No. 02-215-20

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance Statement of accuracy, ISO9001
Includes Polypropylene case
Classification ASTM Class 4
Quantity Each
Packaging Quantity 1
Weight (Metric) 50g
Density 7.95g/cm3 at 20°C
Shape Straight cylinder
Product Type Two piece electronic balance calibration weight
Material 316 Stainless Steel
Mass 50g
Accessories

Accessories for Catalog Number: 0221520

Item Mfr. No. Item Description Price Per Unit Quantity