Please login to your online account to display your discounted pricing

Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 3 with Statement of Accuracy


Precision-crafted of stainless steel, with a highly polished finish

Manufacturer: troemner llc  8316

 View more versions of this product

Catalog No. 02-215-65

  • / Each
 

Description & Specifications

Specifications

Classification ASTM Class 3
Packaging Quantity 1
Includes Polypropylene case
Quantity Each
Density 7.85g/cm3 at 20°C
Certifications/Compliance Statement of accuracy, ISO9001
Shape Cylindrical with groove
Material 303 Stainless Steel
Product Type Two piece electronic balance calibration weight
Mass 5kg
Weight (Metric) 5kg