Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 1 with Statement of Accuracy
Precision-crafted of stainless steel, with a highly polished finish
Manufacturer:  Fisher Scientific 8152F
 View more versions of this product
Catalog No. 02-215-17C
Specifications Description & Specifications
Specifications
Two piece electronic balance calibration weight
ASTM Class 1
50g
50g
Polypropylene case
1
Statement of accuracy, ISO9001
316 Stainless Steel
7.95g/cm3 at 20°C
Straight cylinder
Each
Accessories