Please login to your online account to display your discounted pricing

Troemner Stainless Steel Class 7 Individual Weights


Economical and designed for educational use.

Manufacturer: fisher science education  61013S

 View more versions of this product

Catalog No. S01370

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 1
Certifications/Compliance Statement of accuracy, ISO9001
Classification ASTM Class 7
Shape Cylindrical
Mass 1g
Quantity Each
Weight (Metric) 1g
Density 7.85g/cm3 at 20°C
Product Type Individual metric economical weight
Material 303 Stainless Steel