Troemner Stainless Steel Class 7 Individual Weights
Economical and designed for educational use.
Manufacturer:  Fisher Science Education 61025S
 View more versions of this product
Catalog No. S40268A
Specifications
Description
Weight, Individual; Troemner; Metric; Economical; Stainless Steel; Adjusted to tolerances specified by ANSI/ASTM; Class 7; Nominal mass: 200g
Adjusted to tolerance specified by ANSI/ASTM Class 7.
  • Fractional masses of 200mg and over are made of stainless steel
  • Fractional masses below 200mg are made of aluminum
Description & Specifications
Specifications
Individual metric economical weight with adjusting cavity
ASTM Class 7
200g
200g
Each
Statement of accuracy, ISO9001
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical
1