Please login to your online account to display your discounted pricing

Troemner Stainless Steel Class 7 Individual Weights


Economical and designed for educational use.

Manufacturer: fisher science education  61026S

 View more versions of this product

Catalog No. S63470A

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 1
Quantity Each
Shape Cylindrical
Certifications/Compliance Statement of accuracy, ISO9001
Classification ASTM Class 7
Mass 2000g
Weight (Metric) 2kg
Density 7.85g/cm3 at 20°C
Material 303 Stainless Steel
Product Type Individual metric economical weight with adjusting cavity