Troemner™ Slotted Stainless-Steel Individual Weights
Designed to stack on weight hanger.
Manufacturer:  Troemner™ 53010S
 View more versions of this product
Catalog No. S40385A
Description
Description
  • Adjusted to tolerance specified by ANSI™/ASTM™ Class 7
Specifications
Specifications
Metric slotted weight
Stainless Steel
7.85g/cm3 at 20°C
21mg
4.11cm
ANSI/ASTM Class 7
10g
10g
1.62 in.
Each
Documents