Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, UltraClass with Traceable Certificate


Highly polished kilogram and gram weights

Manufacturer: fisher scientific  70130T

 View more versions of this product

Catalog No. 02-225-537

  • / Each
 

Description & Specifications

Specifications

Product Type Metric analytical weight with handling knob
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass
Material Alloy 8 Stainless Steel
Weight (Metric) 1kg
Density 8.03g/cm3 at 20°C
Mass 1kg
Shape Cylindrical with knob
Includes Polycarbonate case
Quantity Each