Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate
Designed for your most demanding weighing applications
Manufacturer:  Fisher Scientific 70224W
 View more versions of this product
Catalog No. 02-225-507
Specifications Description & Specifications
Specifications
Metric analytical weight with handling knob
ASTM Class 4
5g
5g
Polycarbonate case
1
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with handling knob
Each