Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate


Designed for your most demanding weighing applications

Manufacturer: fisher scientific  70374W

 View more versions of this product

Catalog No. 02-225-533

  • / Each
 

Description & Specifications

Specifications

Density 2.7g/cm3 at 20°C
Product Type Metric analytical sheet weight with one side turned up
Material 3003-H14 Aluminum
Includes Polycarbonate case
Classification ASTM Class 4
Quantity Each
Packaging Quantity 1
Weight (Metric) 1mg
Certifications/Compliance NVLAP, ISO9001
Shape Flat with one side turned up
Mass 1mg