Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate


Designed for your most demanding weighing applications

Manufacturer: fisher scientific  70294W

 View more versions of this product

Catalog No. 02-225-516

  • / Each
 

Description & Specifications

Specifications

Product Type Metric analytical sheet weight with one side turned up
Certifications/Compliance NVLAP, ISO9001
Includes Polycarbonate case
Classification ASTM Class 4
Quantity Each
Packaging Quantity 1
Material 304 Stainless Steel
Weight (Metric) 100mg
Density 7.95g/cm3 at 20°C
Mass 100mg
Shape Flat with one side turned up