Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate


Designed for your most demanding weighing applications

Manufacturer: fisher scientific  70284W

 View more versions of this product

Catalog No. 02-225-515

  • / Each
 

Description & Specifications

Specifications

Product Type Metric analytical sheet weight with one side turned up
Certifications/Compliance NVLAP, ISO9001
Classification ASTM Class 4
Material 304 Stainless Steel
Weight (Metric) 200mg
Density 7.95g/cm3 at 20°C
Mass 200mg
Shape Flat with one side turned up
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1