Troemner™ Individual Analytical Precision Weights, Class 3 with Statement of Accuracy
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer:  Fisher Scientific 70293F
 View more versions of this product
Catalog No. 02-215-30D
Specifications Description & Specifications
Specifications
Each
1
304 Stainless Steel
Polycarbonate case
100mg
Flat with one end turned up for easy handling
ASTM Class 3
7.95g/cm3 at 20°C
One piece metric analytical sheet weight with one side turned up
Statement of accuracy, ISO9001
100mg
Accessories