Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 3 with NVLAP Certificate


Designed for your most demanding weighing applications

Manufacturer: troemner llc  70303W

 View more versions of this product

Catalog No. 02-215-30CC

  • / Each
 

Description & Specifications

Specifications

Product Type One-piece metric analytical sheet weight
Certifications/Compliance NVLAP, ISO9001
Classification ASTM Class 3
Material 304 Stainless Steel
Weight (Metric) 50mg
Density 7.95g/cm3 at 20°C
Mass 50mg
Shape Flat with one end turned up for easy handling
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1