Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 3 with NVLAP Certificate


Designed for your most demanding weighing applications

Manufacturer: fisher scientific  70323W

 View more versions of this product

Catalog No. 02-225-522

  • / Each
 

Description & Specifications

Specifications

Product Type One-piece metric analytical sheet weight
Certifications/Compliance NVLAP, ISO9001
Includes Polycarbonate case
Classification ASTM Class 3
Quantity Each
Material 304 Stainless Steel
Weight (Metric) 20mg
Density 7.95g/cm3 at 20°C
Mass 20mg
Shape Flat with one end turned up for easy handling