Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 2 with Statement of Accuracy


Designed for your most demanding weighing applications

Manufacturer: troemner llc  70362F

 View more versions of this product

Catalog No. 02-217-4B

  • / Each
 

Description & Specifications

Specifications

Product Type One piece metric analytical sheet weight
Certifications/Compliance Statement of accuracy, ISO9001
Includes Polycarbonate case
Classification ASTM Class 2
Quantity Each
Packaging Quantity 1
Material 3003-H14 Aluminum
Weight (Metric) 2mg
Density 2.7g/cm3 at 20°C
Mass 2mg
Shape Flat with one end turned up for easy handling
Accessories

Accessories for Catalog Number: 022174B

Item Mfr. No. Item Description Price Per Unit Quantity