Please login to your online account to display your discounted pricing

Troemner Individual Analytical Precision Weights, Class 2 with NVLAP Certificate


Designed for your most demanding weighing applications

Manufacturer: fisher scientific  70182W

 View more versions of this product

Catalog No. 02-225-502

  • / Each
 

Description & Specifications

Specifications

Product Type Metric analytical weight with handling knob
Certifications/Compliance NVLAP, ISO9001
Classification ASTM Class 2
Material 316 Stainless Steel
Weight (Metric) 50g
Density 7.95g/cm3 at 20°C
Mass 50g
Shape Cylindrical with handling knob
Includes Polycarbonate case
Quantity Each